C-Leg Back-to-Back Stand-Alone Laboratory Benches, ESD, Production Basics
Supplier: Production Basics
Certificates
About this item
This package includes a double-sided frame for free-standing workbench configurations with opposite ESD worksurfaces.
- Appropriate for all applications
- Measures within static dissipative range of 10⁶ to 10⁹
Open, modular design allows for endless accessory possibilities and multi-station configurations. Finished with rounded corners and protective T-mold edge.
Ordering information: Specify a frame and laminate color otherwise will be default blue frame/gray ESD laminate. Frame available in light neutral/almond, blue, white, gray and black. Worksurface available in white, gray and beige. The included 162.6 cm (64") high integrated double-sided back frames are ready to accept accessories. Accessories are sold separately. Order same width accessory as workstation width.
Delivery: Includes (1) double-sided frame, (4) C-leg, adjustable, (2) ESD worksurface, (4) support rail, (8) leveling feet, (1) parts and hardware packet.
Frequently Bought Together