C-Leg Add-on Laboratory Benches, ESD, Production Basics
Supplier: Production Basics
Certificates
About this item
Add an additional ESD workstation to a C-Leg Lab Bench to begin the linear configuration
- Measures within static dissipative range of 10⁶ - 10⁹
Finished with rounded corners and protective T-mold edge.
Ordering information: Requires a C-Leg Stand-Alone Laboratory Bench of same depth, sold separately. When purchased at same time as C-Leg Stand-Alone, the colors will be made to match. If purchasing at separate time, specify a Frame and Laminate color otherwise will be default Blue Frame/Grey ESD Laminate. Frame available in Light Neutral/Almond, Blue, White, Grey and Black. Worksurface available in White, Grey and Beige. The C-Leg included 162.6 cm (64") high integrated back frame is ready to accept accessories. Accessories are sold separately. Order same width accessory as workstation width.
Delivery: Includes (1) single-sided frame, (1) 2C-leg, adjustable, (1) ESD worksurface, (3) support rail, (1) ground bolt and wire, (4) leveling feet, (1) parts and hardware packet.
Frequently Bought Together