Order Entry
United States
ContactUsLinkComponent
Anti-ICAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 15.2]
Anti-ICAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 15.2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ICAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 15.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-514
  • 10050-516
  • Antigen Symbol:
  • ICAM1
  • ICAM1
  • Antigen Name:
  • Intercellular Adhesion molecule 1
  • Intercellular Adhesion molecule 1
  • Antigen Synonyms:
  • ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
  • ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 15.2
  • 15.2
  • Reactivity:
  • Pig, Human
  • Pig, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3383
  • 3383
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together