Order Entry
United States
ContactUsLinkComponent
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-404
  • 10050-406
  • Antigen Symbol:
  • CD3E
  • CD3E
  • Antigen Name:
  • CD3 Molecule Epsilon
  • CD3 Molecule Epsilon
  • Antigen Synonyms:
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • OKT3
  • OKT3
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together