Order Entry
United States
Orders LinkContactUsLinkComponent
 

Keyboard Holder, Production Basics

Supplier:   Production Basics
Keyboard Holder, Production Basics
Supplier:  Production Basics
Certificates
Keyboard Holder, Production Basics
Keyboard Holder, Production Basics

2 Options Available Below

Product Details & Documents

Epoxy powder coated steel holder designed to keep keyboard easily accessible and provides a clutter free worksurface.

  • Keyboard drawer with mouse tray, mounts under the worksurface of any table with a flat mounting area
  • Retractable with built-in wrist rest and 20.3 cm (8") square mouse tray
  • Holds up to 10 lbs.

Compliments any production basics C-Leg lab bench, RTW lab bench, easy lift dual-leg, PedSys lab bench, easy lift 4-post lab bench or use with any generic table.

Weight: 12 lbs.

Ordering information: Standard accessory color is black. Other standard frame color (light neutral/almond, blue, white, or gray) available by request. Accessory is ready to mount under worksurface of any bench.

Product Family Options

Product Information

  • Color
    Availability
    Price
  • 11005-440
    Keyboard and mouse tray
    Keyboard and mouse tray
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $214.46
    $214.46
    New Customer Promotional Price
    Each
       $214.46
     

    Supplier Number: 8681
  • 76239-112
    Keyboard drawer with mouse holder
    Keyboard drawer with mouse holder
    Black
    Log in for availability
    Log in for availability
    Each
    Each - $377.70
    $377.70
    Each
       $377.70

    Supplier Number: PB-8681
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.