Overhead Light, T8, without Mounting Arms, Production Basics
Illuminate your lab bench with Overhead Lighting.
- Overhead Arms not included
- LED tubes included (3)
- Compatible with TYPE B single-end LED tubes ONLY.
- Do not use florescent or other types of LED tubes.
- All units feature energy efficient electronic ballasts to ensure flicker free operation
- Triple-bulb fluorescent lighting units ease operator eye strain while increasing available light at the worksurface
For a replacement light body only or add on to existing Overhead Arms. Use with any Production Basics C-Leg Lab Bench and combine with uprights to RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench, Basic Organizer or Easy Lift 4-post Lab Bench
: Standard accessory color is Black. Other standard frame color (Light Neutral/Almond, Blue, White, or Grey) available by request. This light main body only does not include mounting arms and would mount on existing arms at customer's onsite location.
Frequently Bought Together