UVP Stable Ozone Generator, Analytik Jena
Supplier: Analytik Jena US
Certificates
About this item
Ozone generators produce a continuous flow of ozone as well as a stable source for water purification and air pollution studies
The stable ozone generator utilizes the photochemical reaction of oxygen under shortwave UV to produce a continuous flow of ozone. Generators are a stable source for instrument development and an oxidant source for aging and oxidant testing of materials. These units incorporate Pen-Ray Lamps which are inserted into the generator box and can be removed and used in other applications.
Designed for calibration of field and laboratory ozone analyzers in water purification and air pollution applications.
Ordering information: A power supply that feeds reliable, stable power to the lamps providing uniform ozone generation is included with all models.
Frequently Bought Together