ContactUsLinkComponent
Periscope
Periscope
Catalog # 470117-800
undefined
Periscope
Catalog # 470117-800
Supplier Number:  PSCOPE
Specifications
  • Description:
    Periscope
  • Cat. No.:
    470117-800
  • Supplier no.:
    PSCOPE
Specifications
About this item
Demonstrate how periscopes in submarines work.
  • Sturdy design
  • Mirrors securely held on each end
Inspired by naval periscopes used on submarines, our handheld periscope is designed to show students the reflective qualities of light.  Our version consists of an outer case, with mirrors at each end set parallel to each other at a 45-degree angle.