ContactUsLinkComponent
VWR® PCR Storage Rack
VWR® PCR Storage Rack
Catalog # 83009-690
Supplier:  VWR International
   Sustainable Packaging
   Low Manufacturing Impact
undefined
VWR® PCR Storage Rack
Catalog # 83009-690
Supplier:  VWR International
   Sustainable Packaging
   Low Manufacturing Impact
Specifications
  • Description:
    PCR Storage Rack
  • Environmentally Preferable:
    Sustainable Packaging,
    Low Manufacturing Impact
  • Cat. No.:
    83009-690
Specifications
About this item
The polypropylene 96-well PCR storage rack is perfect for storing samples before cycling or working with completed procedures.
  • Racks hold tubes, strips, or plates that have a separate snap-on lid
  • Autoclavable and freezable
Designed for preparing and storing thin-walled PCR tubes and strip tubes before or after PCR sample processing applications.
Ordering information: Available in assortment pack of five colors: red, orange, yellow, green and blue.