Mouse IgG2b Isotype Control [clone: MPC-11] (redFluor® 710)
Catalog # 10141-258
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Control
- Antigen Name:Mitochondrial pyruvate carrier 11
- Clone:MPC-11
- Conjugation:redFluor® 710
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10141-258
- Supplier no.:80-4732-U100
Specifications
About this item
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Type: Control
Antigen: MPC11
Clonality:
Clone: MPC-11
Conjugation: redFluor® 710
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: