Order Entry
United States
ContactUsLinkComponent
Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Monoclonal Antibody [clone: M5/114.15.2] (PE)
Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Monoclonal Antibody [clone: M5/114.15.2] (PE)
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Monoclonal Antibody [clone: M5/114.15.2] (PE)
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-340
  • 10051-338
  • Antigen Symbol:
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • Antigen Name:
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • Antigen Synonyms:
  • MHC II|IA/IE|IA|IE|I-A/E
  • MHC II|IA/IE|IA|IE|I-A/E
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • M5/114.15.2
  • M5/114.15.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 14961/14969
  • 14961/14969
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together