Mouse IgG2b Monoclonal Isotype Control [clone: MPC-11] (FITC (Fluorescein Isothiocyanate))
10050-988
- Antibody Type:Control
- Antigen Name:Mitochondrial pyruvate carrier 11
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10050-988
- Supplier no.:35-4732-U100
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Type: Control
Antigen: MPC11
Clonality: Monoclonal
Clone: MPC-11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: