Rat IgG2a Isotype Control [clone: 2A3]
Catalog # 10141-176
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Control
- Antigen Name:UDP-glucuronosyltransferase 2A3
- Clone:2A3
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested Applications:Functional Assay
- Cat. No.:10141-176
- Supplier no.:40-4321-U100
Specifications
About this item
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.
Type: Control
Antigen: 2A3
Clonality:
Clone: 2A3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: