Rat IgG2a Monoclonal Isotype Control [clone: 2A3] (FITC (Fluorescein Isothiocyanate))
Catalog # 10050-982
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Control
- Antigen Name:UDP-glucuronosyltransferase 2A3
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10050-982
- Supplier no.:35-4321-U100
Specifications
About this item
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.
Type: Control
Antigen: 2A3
Clonality: Monoclonal
Clone: 2A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: