Storage Boxes with Locks, Mitchell Plastics
Supplier: Avantor
Certificates
About this item
Store contents from unauthorized users in these various storage boxes
"Also available are storage boxes for the wall with an electronic touch pad or an easily reprogrammable combination lock. All boxes are made of 3/8" thick acrylic.
Refrigerator storage boxes have four holes for securing in a refrigerator or cabinet. Refrigerator storage boxes are available with easily reprogrammable combination locks or with a lockable pull out drawer. They can withstand temperatures as low as -70°C.
Frequently Bought Together