Order Entry
United States
ContactUsLinkComponent
 
Anti-IL2RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BC96]
 
undefined
Anti-IL2RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BC96]

 

  • Catalog No:
  • 10050-474
  • 10050-476
  • Antigen Symbol:
  • IL2RA
  • IL2RA
  • Antigen Name:
  • Interleukin 2 Receptor, Alpha
  • Interleukin 2 Receptor, Alpha
  • Antigen Synonyms:
  • TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • BC96
  • BC96
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3559
  • 3559
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together