Order Entry
United States
ContactUsLinkComponent
 
Anti-CD28 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: CD28.2]
 
undefined
Anti-CD28 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: CD28.2]

 

  • Catalog No:
  • 10051-588
  • 10051-590
  • Antigen Symbol:
  • CD28
  • CD28
  • Antigen Name:
  • CD28 Molecule
  • CD28 Molecule
  • Antigen Synonyms:
  • Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
  • Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • CD28.2
  • CD28.2
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 940
  • 940
  • Isotype:
  • IgG1
  • IgG1
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together