Order Entry
United States
ContactUsLinkComponent
Sodium hypochlorite (6% Cl₂) in aqueous solution
Sodium hypochlorite (6% Cl₂) in aqueous solution
Supplier:  Ricca Chemical
Certificates
undefined
Sodium hypochlorite (6% Cl₂) in aqueous solution
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Assay:
    6 ± 0.06 % (w/w)
  • NIST SRM:
    136

Specification Test Results

Related Information

Frequently Bought Together