Sodium hydroxide 33.3% (w/w) in aqueous solution
Catalog # RC7278.3-1
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly natural
- Size:4 L
- Cat. No.:RC7278.3-1
- Supplier no.:7278.3-1
Specifications
Specification Test Results
- Assay:33.3 ± 1 % (w/w)
- NIST SRM:723
Specification Test Results
About this item
:
:40 g/mol
UN:1824
ADR:8,II
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548