Reagent for chlorine (residual) by DPD method, Potassium Permanganate 891 mg/L (1000 ppm mg/L as Cl₂), Ricca Chemical Company
RCR6354300500C
Small Business Enterprise
:
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Pack type:Amber Glass
- Size:500 mL
- Cat. No.:RCR6354300500C
- Supplier no.:R6354300-500C
Suitable for use in APHA 3500-Cl G as Reagent 4a2, Potassium Permanganate Stock