Potassium hydroxide 0.0100 N (N/100) in 2-propanol
RC6253-32
Small Business Enterprise
:
- Pack type:Poly natural
- Size:1 L
- Cat. No.:RC6253-32
- Supplier no.:6253-32
- Assay:0.01 ± 0.00005 N at 20 °C
:
:56.11 g/mol
:2924
:3,II
:1310-58-3
:KOH
:MFCD00003553