Order Entry
United States
ContactUsLinkComponent
 
Perchloric acid 8% (w/v) in aqueous solution
Catalog # RCR5512800100A
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Perchloric acid 8% (w/v) in aqueous solution
Catalog # RCR5512800100A
Supplier:  Ricca Chemical
Supplier Number:  R5512800-100A
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly natural
  • Size:
    100 ml
  • Cat. No.:
    RCR5512800100A
  • Supplier no.:
    R5512800-100A

Specifications

Specification Test Results

  • Assay:
    8 ± 0.01 % (w/v)
  • NIST SRM:
    84

Specification Test Results

About this item

 :
 :100.46 g/mol
UN:1802
ADR:8,II
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325