Nitrite Standard, 1 ml = 1 mg N, 1000 ppm N as Nitrite (3285 ppm NO₂⁻), Ricca Chemical Company
Catalog # RCR5444900120C
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 ppm
- Pack type:Glass bottle, amber
- Size:120 ml
- Cat. No.:RCR5444900120C
- Contents:KNO₂ in H₂O
- Supplier no.:R5444900-120C