Hydrochloric acid (1 + 9) (v/v) 10% (v/v) in aqueous solution
Catalog # RC3570-2.5
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Cubitainer®
- Size:10 L
- Cat. No.:RC3570-2.5
- Supplier no.:3570-2.5
Specifications
Specification Test Results
- Assay:10 ± 0.08 % (v/v)
- NIST SRM:84
Specification Test Results
About this item
:
ADR:8,II
:36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324