ContactUsLinkComponent
DPD Indicator Solution, for Residual Chlorine Analysis
DPD Indicator Solution, for Residual Chlorine Analysis
Catalog # RC2655-1
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
DPD Indicator Solution, for Residual Chlorine Analysis
Catalog # RC2655-1
Supplier:  Ricca Chemical
Supplier Number:  2655-1
Small Business Enterprise
CAS Number:  
Specifications
  • Pack type:
    Amber Glass
  • Size:
    4 L
  • Cat. No.:
    RC2655-1
  • Supplier no.:
    2655-1
Specifications
About this item
Suitable for use in APHA 4500-Cl G and EPA Method 330.5 for Residual Chlorine Analysis