Cell Detachment Solution, MP Biomedicals
Catalog # IC1000449
Supplier: MP Biomedicals
CAS Number:
Specifications
- Size:100ml
- Cat. No.:IC1000449
- Supplier no.:1000449
- Activity:≥200 u/ml
Specifications
About this item
Accutase® solution is a novel trypsin replacement that exhibits both protease and collagenolytic activities.
- Maintains most cell surface antigens
- Contains proteolytic and collagenolytic enzymes
- Suitable for Cell Culture
Accutase® Cell Detachment Solution is effective in detaching primary fibroblasts, endothelial cells, neurons, tumor cell lines, and insect cells. Performs exceptionally well in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up.
Solution: Supplied in 0.2 g/L KCl, 8 g/L NaCl, 0.2 g/L KH2PO4, 1.15 g/L Na2HPO4, 0.5 mM EDTA 4 Na, 3 mg/L Phenol Red Performance (Cell Culture Tested)