About this item
Defined fetal bovine serum (FBS) is the highest quality of FBS available and is very widely used by those cell culturist who have a concern for viral contaminants and require an extensive biochemical profile. Defined FBS is filtered through multiple 40 nm (0.04 µm) pore-size related filters, which are the most retentive filters used in commercial FBS production. Over 50 components are analysed and the results are included on the biochemical assay.
- Filtered through serial 40 nm pore-size rated filters
- Maximize cell culture growth
- U.S. sourced
Maximize cell culture growth with U.S.-sourced Cytiva HyClone Defined Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 16777-530
- 82013-568
- 16777-006
- 16777-002
- 16750-030
- Storage Temperature:
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
Specifications
Related Information
Certificates
Search by Lot NumberCytiva BioXtra and Pall Life Sciences Products CA Cytiva BioXtra and Pall Life Sciences Products US
Promotion Material
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Cytiva HyClone Media and Supplements HyClone™ Cell Culture Serum products