GeneMate Western Blot Boxes, Blotting Containers, MTC Bio
Supplier: GeneMate
Certificates
About this item
These western blot boxes are made of polystyrene and are ideal for halves, wholes, strips or minis.
- Reduce antibody usage
- Features 90° angle corners
These boxes feature easy-to-open hinged lid which reduces evaporation.
Frequently Bought Together