Orders LinkContactUsLinkComponent
Rabbit Dissection
Rabbit Dissection
Rabbit Dissection Embedment
Catalog # 470150-518
Supplier #:  T608
Rabbit Dissection
Catalog # 470150-518
Rabbit Dissection
Rabbit Dissection
Rabbit Dissection Embedment
Price
$145.95
Log in for availability
Each
Product Details & Documents
This kit looks at the rabbit dissection
There is one specimen block included (10x4 1/8 x2 1/8in.). The specimen is preserved and encased in clear resin material. It can be observed from every side to see details. The specimen is a rabbit (Lepus sinensis). Teacher information guide, student activity sheet, and answer key are included. Size of product is 12.6x6.0x4.0in.
Note: Ward's preserved rabbits are a byproduct of the food industry. Rabbits are not raised specifically for dissection. These animals are "recycled" for educational use.
Specifications
  • Description:
    Rabbit Dissection Embedment