Order Entry
United States
ContactUsLinkComponent
 
Anti-LY76 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TER-119]
 
undefined
Anti-LY76 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TER-119]

 

  • Catalog No:
  • 10051-020
  • 10051-022
  • 10051-018
  • Antigen Symbol:
  • LY76
  • LY76
  • LY76
  • Antigen Name:
  • Lymphocyte Antigen 76
  • Lymphocyte Antigen 76
  • Lymphocyte Antigen 76
  • Antigen Synonyms:
  • Ter119|TER-119
  • Ter119|TER-119
  • Ter119|TER-119
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • TER-119
  • TER-119
  • TER-119
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 104231
  • 104231
  • 104231
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Yes
  • Storage Temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together