Order Entry
United States
ContactUsLinkComponent
Rat IgG1 Isotype Control [clone: HRPN]
Rat IgG1 Isotype Control [clone: HRPN]
Catalog # 10141-214
Supplier:  Tonbo Biosciences
undefined
Rat IgG1 Isotype Control [clone: HRPN]
Catalog # 10141-214
Supplier:  Tonbo Biosciences
Supplier Number:  70-4301-U100

Specifications

  • Antibody Type:
    Control
  • Clone:
    HRPN
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Size:
    100 µg
  • Storage Temperature:
    Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested Applications:
    Immuhohistochemistry,
  • Cat. No.:
    10141-214
  • Supplier no.:
    70-4301-U100

Specifications

About this item

The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: