Rat IgG1 Monoclonal Isotype Control [clone: HRPN] (PE (Phycoerythrin)-Cy7®)
10051-524
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10051-524
- Supplier no.:60-4301-U100
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: HRPN
Clonality: Monoclonal
Clone: HRPN
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: