Rat IgG1 Isotype Control [clone: HRPN] (FITC (Fluorescein Isothiocyanate))
10141-158
- Antibody Type:Control
- Clone:HRPN
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Size:100 µg
- Storage Temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10141-158
- Supplier no.:35-4301-U100
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: