Rat IgG1 Monoclonal Isotype Control [clone: HRPN] (APC (Allophycocyanin))
Catalog # 10050-604
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10050-604
- Supplier no.:20-4301-U100
Specifications
About this item
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: HRPN
Clonality: Monoclonal
Clone: HRPN
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: