About this item
The OKT4 antibody reacts with human CD4, a 59 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.
The OKT4 antibody is widely used as a phenotypic marker for CD4 expression. It is cross-reactive with CD4 in several non-human species, including Chimpanzee, Cynomolgus and Rhesus. This antibody recognizes a different epitope, and thus does not block binding of, the alternative Anti-Human CD4 antibody clone RPA-T4 (Reinherz EL, et al. 1979. Proc. Natl. Acad. Sci. 76:4061-4065).
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Specifications
- Catalog No:
- 10050-422
- 10050-418
- 10050-420
- Antigen Symbol:
- CD4
- CD4
- CD4
- Antigen Name:
- CD4 molecule
- CD4 molecule
- CD4 molecule
- Antigen Synonyms:
- L3T4|p55|CD4mut|Ly-4|W3/25
- L3T4|p55|CD4mut|Ly-4|W3/25
- L3T4|p55|CD4mut|Ly-4|W3/25
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- OKT4
- OKT4
- OKT4
- Reactivity:
- Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 920
- 920
- 920
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- Flow Cytometry:
- Yes
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Yes
- Storage Temperature:
- Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested Applications:
- Flow Cytometry
- Flow Cytometry
- Flow Cytometry