Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 10141-206
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:C1.18.4
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10141-206
- Supplier no.:65-4724-U100
Specifications
About this item
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: Control
Antigen: C1.18.4
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0