Rat IgG2b Monoclonal Isotype Control [clone: LTF-2] (PE (Phycoerythrin))
Catalog # 10051-326
Supplier: Tonbo Biosciences
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:LTF-2
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Size:100 µg
- Neutralisation:Yes
- Storage Temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested Applications:Flow Cytometry
- Cat. No.:10051-326
- Supplier no.:50-4031-U100
Specifications
About this item
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.
Type: Control
Antigen: LTF-2
Clonality: Monoclonal
Clone: LTF-2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: