Order Entry
United States
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: RM4-5]
Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: RM4-5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD4 Rat Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: RM4-5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-676
  • 10050-678
  • Antigen Symbol:
  • CD4
  • CD4
  • Antigen Name:
  • CD4 molecule
  • CD4 molecule
  • Antigen Synonyms:
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • RM4-5
  • RM4-5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage Temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested Applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together