Inductor
470112-964
- Description:Inductor
- Environmentally Preferable:
- Cat. No.:470112-964
- Supplier no.:IND
This air-core inductor is made from 20 gauge copper wire.
It has an inductance of 5 mH and low resistance. A metal core can be added to increase inductance.