Vernier® Green Diffraction Laser
Vernier green diffraction laser
Catalog # 470112-944
Supplier: Vernier Software & Technology
Supplier #: GDL-DAK
Jump to:
Product Details & Documents
The Green Diffraction Laser is an optional accessory for the Diffraction Apparatus.
Operating at 532 nm, it allows students to see and measure how diffraction patterns depend on light wavelength.
Specifications
- Description:Vernier green diffraction laser