Order Entry
United States
Orders LinkContactUsLinkComponent
PermaRed/AP Auto, Diagnostic BioSystems

PermaRed/AP Auto, Diagnostic BioSystems

PermaRed/AP Auto, 110 mL
Catalog # 10098-720
Supplier #:  K049-AUTO-110
PermaRed/AP Auto, Diagnostic BioSystems
Catalog # 10098-720
CAS Number:  
PermaRed/AP Auto, Diagnostic BioSystems

PermaRed/AP Auto, 110 mL

CAS Number:  
Price
$545.09
Log in for availability
Each (110ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PermaRed/AP-Auto produces a red color, ranging from pink to brilliant dark red, depending on the signal intensity

PermaRed/AP-Auto has been formulated for on-slide mixing. The instrument should be programmed to apply the substrate-buffer to the slide followed by application of the chromogen reagent. The substrate-buffer and chromogen components are stable in their separate formats. They can be loaded into the reagent rack at the beginning of the staining run with the other reagents, for walk away convenience.

It is insoluble in organic solvents; therefore sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted.

Specifications

Specifications

  • Description:
    PermaRed/AP Auto
  • Size:
    110 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.