ContactUsLinkComponent
Vernier® Resistor Board
Vernier® Resistor Board
Catalog # 470116-634
undefined
Vernier® Resistor Board
Catalog # 470116-634
Supplier Number:  VES-RB
Specifications
  • Description:
    Vernier Resistor Board
  • Cat. No.:
    470116-634
  • Supplier no.:
    VES-RB
Specifications
About this item
The Vernier Resistor Board has seven 1 W 5% resistors mounted on a circuit board.
Test clip holes on either side of the resistors allow for easy connections to clips on wire leads.