ContactUsLinkComponent
Motion Detector Clamp
Motion Detector Clamp
Catalog # 470113-950
undefined
Motion Detector Clamp
Catalog # 470113-950
Supplier Number:  MD-CLAMP
Specifications
  • Description:
    Motion Detector Clamp
  • Cat. No.:
    470113-950
  • Supplier no.:
    MD-CLAMP
Specifications
About this item
The Motion Detector Clamp screws into the back of the Motion Detector, and then can be clamped to a variety of objects such as table tops, ring stands, etc.