ContactUsLinkComponent
Accessories for Vernier® Rotary Motion Sensor, Vernier Software & TECH SE
Accessories for Vernier® Rotary Motion Sensor, Vernier Software & TECH SE
Catalog # 470112-968
undefined
Accessories for Vernier® Rotary Motion Sensor, Vernier Software & TECH SE
Catalog # 470112-968
Supplier Number:  AK-RMV
Specifications
  • Description:
    Rotational motion accessory kit
  • Cat. No.:
    470112-968
  • Supplier no.:
    AK-RMV
Specifications
About this item
Investigate the motion of a pendulum.
  • Used with the Rotary Motion Sensor
This accessory kit is used with the Rotary Motion Sensor to study the motion of a physical pendulum; the rotational inertia of disks, rings, and point masses; and the conservation angular momentum.
Two 3-1/2 inch diameter, 1/4 inch thick solid aluminum disks
Steel disk with 1-1/8 inch central hole
Plastic hub to hold the steel disk on the 3-step pulley
15 inch hollow aluminum rod
Ultra Pulley
Ultra Pulley Swivel Mount, which allows the pulley to be mounted on the Rotary Motion Sensor
2 Masses with locking screws
For educational use only.