Corning® 1536 Well Cyclic Olefin Copolymer (COC) Microplate, Corning
Supplier: Corning
Certificates
About this item
Cyclic olefin copolymer (COC). Superior flatness of the plates minimizes variability within a plate and between plates. The barcoded plates come with with raised well bottoms for higher sensitivity.
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- No logo or lettering for improved heat sealing and ease of liquid handling
- Working volume is up to 10 µl
- Flat bottom wells are compatible with automated dispensers
Specifications
- Catalog No:
- 89423-300
- 89423-302
- 89423-304
- 89423-306
- 89423-308
- 89423-312
- 89423-314
- 89423-316
- 89423-296
- 89423-298
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk