Synthware Solvent Still with Built-In Condenser, Kemtech America
60004-770
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- [ST] joint:24/40
- Capacity:125 ml
- Description:Synthware solvent still with built-in condenser
- Cat. No.:60004-770
- Supplier no.:S144125
Grease-free design.
- Built in condenser
- Standard 24/40 ground glass joint
- Stopper and cap included
For excellent solvent and chemical resistance, the still is equipped with two 0-4mm high vacuum valves that have PTFE covers over the two Viton® O-rings on the shaft, and one integral PTFE O-ring on the tip, without exposing any material other than PTFE to any solvents. Includes 14/10 threaded joint and PTFE-lined septum for syringe access.