Nunc® 384-Well ShallowWell Standard Height Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Reduce lab expenditures in samples and reagents using Nunc® 384-Well ShallowWell Standard Height Plates
Total well volume is reduced to 25μL. The standard height of the plate allows the use of automation without making adjustments to robotics, plate readers, or liquid handlers. Plates are availabe in clear, black, or white formats for colorimetric, fluorescent, and luminescent assays.
Certifications: All microplates manufactured by Nunc® meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004).
Related Information
Certificates
Search by Lot NumberThermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Frequently Bought Together