Lead, foil
Roll, 5 lbs.
Catalog # 11019-412
Supplier: ALPHA ASSEMBLY SOLUTIONS, INC.
Supplier #: 142733
Jump to:
Product Details & Documents
Lead Foil is typically used in both Fire Assay bullion work and cupellation. For example to wrap bullion samples, or for additional cupellation purposes.
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Merck Index:12,05410
- Boiling Pt:1740 °C (1013 hPa)
- Melting Pt:327 °C
- Density:11.3437 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-100-4
Specifications
Specification Test Results
- Thickness:0.1 mm
Specifications
- Pack type:Roll
- Size:5 lbs.
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



