Bead Anvil, Denco Equipment and Machining
Catalog # CA11019-682
Supplier: DENCO EQUIPMENT & MACHINING CO. CA
Specifications
- Description:Bead Anvil
- Height:50 mm
- Cat. No.:CA11019-682
- Supplier no.:FA120
- Diameter:76 mm
Specifications
About this item
Ideal for flatenning beads, the bead anvil is constructed from shaft steel with a hardened surface.
Ordering information: Contact your Avantor sales representative for more information.