B-Phycoerythrin (BPE), Columbia Biosciences
Catalog # 10065-812
Supplier: Columbia Biosciences
CAS Number:
Specifications
- Description:B-Phycoerythrin (BPE)
- Size:1 mg
- Storage Temperature:Store at 4° C
- Application:Flow Cytometry, Cell-Based Assays, Microarrays and Microplate Applications
- Ex/em:545/572 nm
- Cat. No.:10065-812
- Dye:SureLight® B-Phycoerythrin (B-PE)
- Supplier no.:D4-000
Specifications
About this item
Because of its high quantum yield, B-PE is considered the world’s brightest fluorophore
It is compatible with commonly available lasers and gives exceptional results in flow cytometry, Luminex® and immunofluorescent staining. B-PE is also less “sticky” than common synthetic fluorophores and therefore gives less background interference.